Invitrogen
Manufacturer Code:MA116731
Catalog # AMPA6731
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 464.6 |
Immunogen | Purified NaK-ATPase from rabbit renal outer medulla. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: (Na+, K+)-ATPase alpha-subunit; ATP1B MGC1798; ATPase, Na+/K+ transporting, alpha 1 polypeptide; ATPase, Na+K+ transporting, alpha 1; Na(+)/K(+) ATPase alpha-1 subunit; Na+ ,K+ -ATPase alpha subunit; Na+ /K+ ATPase alpha 1 subunit; Na+, K+-ATPase beta-subunit precursor; Na+/K+ -ATPase alpha 1 subunit; Na+/K+ ATPase; Na+/K+ ATPase 1; Na+/K+ transporting alpha 1 polypeptide; Na+K+ ATPase; Na, K-ATPase, alpha-A catalytic polypeptide; Na,K-ATPase alpha-1 subunit; Na,K-ATPase catalytic subunit alpha-A protein; Na/K ATPase alpha 1 subunit; sodium pump 1; Sodium pump subunit alpha-1; sodium-potassium ATPase catalytic subunit alpha-1; sodium-potassium-ATPase, alpha 1 polypeptide; sodium/potassium-ATPase alpha-subunit 1; sodium/potassium-transporting ATPase alpha-1 chain; Sodium/potassium-transporting ATPase subunit alpha-1; unnamed protein product
Gene Aliases: ATP1A1; atp1a1-a; atp1a1-b; atp1a1a.4; Atpa-1; BC010319; K-ATPase; Na+K+ATPase; Nkaa1b
UniProt ID: (Dog) P50997, (Sheep) P04074, (Rabbit) Q9N0Z6, (Human) P05023, (Pig) P05024, (Bovine) Q08DA1, (Rat) Q64609, (Mouse) Q8VDN2
Entrez Gene ID: (Dog) 403992, (Sheep) 443381, (Rabbit) 100302415, (Human) 476, (Pig) 397481, (Bovine) 282144, (Guinea pig) 100192382, (Rat) 24211, (Mouse) 11928, (Western clawed frog) 395044
Molecular Function:
cation transporter
hydrolase
ion channel
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.