Invitrogen
Manufacturer Code:PA522852
Catalog # PIPA522852
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide within the internal region of residues 200-300 of the human ATP13A2 protein. |
Conjugate | HRP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATPase type 13A2; Polyamine-transporting ATPase 13A2; probable cation-transporting ATPase 13A2; RP1-37C10.4 HSA9947 KRPPD PARK9 probable cation-transporting ATPase 13A2 putative ATPase
Gene Aliases: 1110012E06Rik; AA589443; ATP13A2; CLN12; HSA9947; KRPPD; PARK9
UniProt ID: (Human) Q9NQ11, (Mouse) Q9CTG6
Entrez Gene ID: (Human) 23400, (Mouse) 74772
Molecular Function: cation transporter hydrolase ion channel transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.