Novus Biologicals
Manufacturer Code:NB11041486AF532
Catalog # N1041486A32
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to an internal region of the human ATP13A2 protein (within residues 200-300). [Swiss-ProtNo. Q9NQ11] |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
Protein Aliases: ATPase type 13A2; ATPase type 13A2 EC 3.6.3 EC 3.6.3.- EC 3.6.3.5 EC 3.6.3.8 FLJ26510 HSA9947 KRPPD PARK9putative ATPase Parkinson disease (autosomal recessive) 9 (Kufor-Rakeb syndrome) probable cation-transporting ATPase 13A2; Polyamine-transporting ATPase 13A2
Gene Aliases: ATP13A2; CLN12; HSA9947; KRPPD; PARK9
UniProt ID: (Human) Q9NQ11
Entrez Gene ID: (Human) 23400
Molecular Function: cation transporter hydrolase ion channel transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.