Abnova
Manufacturer Code:PAB0697
Catalog # 89-114-349
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to amino acids 195-209 of human ATP13A2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C on dry atmosphere. After reconstitution with deionized water,store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of ATP13A2.
For Research Use Only
Protein Aliases: ATPase type 13A2; FLJ26510; HSA9947; KRPPD; PARK9; Polyamine-transporting ATPase 13A2
Gene Aliases: ATP13A2; CLN12; HSA9947; KRPPD; PARK9
UniProt ID: (Human) Q9NQ11
Entrez Gene ID: (Human) 23400
Molecular Function: cation transporter hydrolase ion channel transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.