Invitrogen
Manufacturer Code:PA558826
Catalog # PIPA558826
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to Human ATP12A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Store at &minus20°C long term. Avoid freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATPase, H+/K+ transporting, nongastric, alpha polypeptide; ATPase, Na+/K+ transporting, alpha polypeptide-like 1; non-gastric H(+)/K(+) ATPase alpha subunit; Non-gastric H(+)/K(+) ATPase subunit alpha; Potassium-transporting ATPase alpha chain 2; Potassium-transporting ATPase alpha chain 2 Non-gastric H(+)/K(+) ATPase subunit alpha Proton pump ATP12A ATP1AL1; Proton pump
Gene Aliases: #945; ATP12A; ATP1AL1; HK
UniProt ID: (Human) P54707
Entrez Gene ID: (Human) 479
Molecular Function:
cation transporter
hydrolase
ion channel
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.