Invitrogen
Manufacturer Code:PA5100678
Catalog # PIPA5100678
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ATP12A corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: #945 ATP12A ATP1AL1 ATPase H+/K+ transporting non-gastric alpha2 subunit ATPase H+K+-transporting alpha 2 ATPase H+/K+ transporting nongastric alpha polypeptide ATPase Na+/K+ transporting alpha polypeptide-like 1 cHKA HK HKalpha2 non-gastric H(+)/K(+) ATPase alpha subunit Non-gastric H(+)/K(+) ATPase subunit alpha potassium-transporting ATPase alpha chain 2 Proton pump; ATPase H+K+-transporting, alpha 2; ATPase, H+/K+ transporting, nongastric, alpha polypeptide; ATPase, Na+/K+ transporting, alpha polypeptide-like 1; non-gastric H(+)/K(+) ATPase alpha subunit; Non-gastric H(+)/K(+) ATPase subunit alpha; Potassium-transporting ATPase alpha chain 2; Proton pump
Gene Aliases: #945; ATP12A; ATP1AL1; cHKA; HK; HKalpha2
UniProt ID: (Human) P54707, (Mouse) Q9Z1W8
Entrez Gene ID: (Human) 479, (Mouse) 192113
Molecular Function: cation transporter hydrolase ion channel transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.