Novus Biologicals
Manufacturer Code:NBP292160100UL
Catalog # NB026211
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 890-980 of human ATP12A (NP_001172014.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP1AL1non-gastric H(+)/K(+) ATPase alpha subunit ATPase H+/K+ transporting nongastric alpha polypeptide ATPase Na+/K+ transporting alpha polypeptide-like 1 EC 3.6.3 EC 3.6.3.10 Non-gastric H(+)/K(+) ATPase subunit alpha potassium-transporting ATPase alpha chain 2 Proton pump; ATPase, H+/K+ transporting, nongastric, alpha polypeptide; ATPase, Na+/K+ transporting, alpha polypeptide-like 1; non-gastric H(+)/K(+) ATPase alpha subunit; Non-gastric H(+)/K(+) ATPase subunit alpha; Potassium-transporting ATPase alpha chain 2; Proton pump
Gene Aliases: #945; ATP12A; ATP1AL1; HK
UniProt ID: (Human) P54707
Entrez Gene ID: (Human) 479
Molecular Function: cation transporter hydrolase ion channel transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.