Invitrogen
Manufacturer Code:PA525361
Catalog # PIPA525361
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 589-616 amino acids from the central region of human ATP11C |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Atpase 11c, p type; ATPase class VI type 11C; ATPase IQ; ATPase, class VI, type 11C; ATPIG ATPIQ phospholipid-transporting ATPase IG probable phospholipid-transporting ATPase IG; P4-ATPase flippase complex alpha subunit ATP11C; Phospholipid-transporting ATPase 11C; Phospholipid-transporting ATPase IG; probable phospholipid-transporting ATPase 11C; probable phospholipid-transporting ATPase IG; putative phospholipid-transporting ATPase 11C
Gene Aliases: A330005H02Rik; AI315324; ATP11C; ATPIG; ATPIQ; I79_007666; Ig
UniProt ID: (Human) Q8NB49, (Mouse) Q9QZW0
Entrez Gene ID: (Human) 286410, (Mouse) 320940, (Chinese hamster) 100756336
Molecular Function: cation transporter hydrolase transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.