Invitrogen
Manufacturer Code:PA520996
Catalog # PIPA520996
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 19 amino acid peptide near the amino terminus of human ATP11B. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATPase class VI type 11B; ATPase IR; ATPase, class VI, type 11B; ATPIF ATPIR ATPase IR probable phospholipid-transporting ATPase IF; P4-ATPase flippase complex alpha subunit ATP11B; Phospholipid-transporting ATPase IF; probable phospholipid-transporting ATPase IF; truncated ATPase 11B protein
Gene Aliases: 1110019I14Rik; ATP11B; ATPIF; ATPIR; KIAA0956; mKIAA0956
UniProt ID: (Human) Q9Y2G3, (Mouse) Q6DFW5
Entrez Gene ID: (Human) 23200, (Mouse) 76295
Molecular Function: cation transporter hydrolase transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.