Invitrogen
Manufacturer Code:PA557324
Catalog # PIPA557324
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human ATP11A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATPase class VI type 11A; ATPase IS; ATPase, class VI, type 11A; P4-ATPase flippase complex alpha subunit ATP11A; phospholipid-translocating ATPase; Phospholipid-transporting ATPase IH; potential phospholipid-transporting ATPase IH; RP11-120K24.1 ATPIH ATPIS phospholipid-translocating ATPase potential phospholipid-transporting ATPase IH probable phospholipid-transporting ATPase IH
Gene Aliases: ATP11A; ATPIH; ATPIS; KIAA1021
UniProt ID: (Human) P98196
Entrez Gene ID: (Human) 23250
Molecular Function: cation transporter hydrolase transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.