Novus Biologicals
Manufacturer Code:NBP214327
Catalog # NBP214327
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: LFPSPILRAKHFDRLTPEERTKALKKWRGAGKMNQVTSKYANQSAGKSGR RPMPGPSAVFAMKSASSCAIEQGNLSLCETALDQGYSETKAFEMAGPS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATPase class V type 10D; ATPase class V type 10D ATPVDtype IV aminophospholipid transporting ATPase EC 3.6.3 EC 3.6.3.1 KIAA1487ATPase class V type 10D probable phospholipid-transporting ATPase VD; ATPase, class V, type 10D; P4-ATPase flippase complex alpha subunit ATP10D; Phospholipid-transporting ATPase VD; type IV aminophospholipid transporting ATPase
Gene Aliases: ATP10D; ATPVD; KIAA1487
UniProt ID: (Human) Q9P241
Entrez Gene ID: (Human) 57205
Molecular Function: cation transporter hydrolase transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.