Invitrogen
Manufacturer Code:PA529497
Catalog # PIPA529497
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 2 and 285 of Human ATP citrate lyase |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACL; ACL ATPCL CLATP ATP citrate synthase ATP-citrate (pro-S-)-lyase ATP-citrate synthase citrate cleavage enzyme; ATP-citrate (pro-S-)-lyase; ATP-citrate synthase; Citrate cleavage enzyme
Gene Aliases: A730098H14Rik; ACL; ACLY; ATPCL; AW538652; CLATP
UniProt ID: (Human) P53396, (Mouse) Q91V92, (Rat) P16638
Entrez Gene ID: (Human) 47, (Mouse) 104112, (Rat) 24159
Molecular Function:
ligase
lyase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.