Abnova
Manufacturer Code:H00023460A01
Catalog # 89-004-564
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | ABCA6 (NP_525023 53 a.a. ~ 149 a.a) partial recombinant protein with GST tag. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant ABCA6.
For Research Use Only
Protein Aliases: ABC transporter ABCA6; ATP-binding cassette A6; ATP-binding cassette sub-family A member 6; ATP-binding cassette, sub-family A (ABC1), member 6; EST155051; FLJ43498
Gene Aliases: ABCA6; EST155051
UniProt ID: (Human) Q8N139
Entrez Gene ID: (Human) 23460
Molecular Function: ATP-binding cassette (ABC) transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.