Abnova
Manufacturer Code:H00000475M10
Catalog # 89-016-258
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3B7 |
Immunogen | ATOX1 (NP_004036 1 a.a. ~ 68 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant ATOX1.
For Research Use Only
Protein Aliases: ATX1; ATX1 antioxidant protein 1 homolog; Copper transport protein ATOX1; HAH1; Metal transport protein ATX1; MGC138453; MGC138455
Gene Aliases: ATOX1; ATX1; HAH1
UniProt ID: (Human) O00244
Entrez Gene ID: (Human) 475
Molecular Function: chaperone transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.