Novus Biologicals
Manufacturer Code:NBP292669100UL
Catalog # NB027207
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-68 of human ATOX1 (NP_004036.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATX1 antioxidant protein 1 homolog; ATX1 ATX1 (antioxidant protein 1 yeast) homolog 1 ATX1 antioxidant protein 1 homolog (yeast) copper transport protein ATOX1 hah1 HAH1 MGC138453 Metal transport protein ATX1 MGC138455; Copper transport protein ATOX1; Metal transport protein ATX1
Gene Aliases: ATOX1; ATX1; HAH1
UniProt ID: (Human) O00244
Entrez Gene ID: (Human) 475
Molecular Function: chaperone transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.