Proteintech
Manufacturer Code:192431AP150UL
Catalog # 50-173-2373
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ATN1 Fusion Protein Ag5835 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATN1 atrophin 1 B37 D12S755E DRPLA HRS NOD; Atrophin-1; dentatorubral pallidoluysian atrophy; Dentatorubral-pallidoluysian atrophy protein; Dentatorubral-pallidoluysian atrophy protein homolog
Gene Aliases: ATN1; Atr1; atrophin-1; B37; D12S755E; DRPLA; HRS; NOD
UniProt ID: (Human) P54259, (Mouse) O35126
Entrez Gene ID: (Human) 1822, (Mouse) 13498
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.