Novus Biologicals
Manufacturer Code:NB100306C
Catalog # NB100306C
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 10H11.E12 |
Immunogen | Synthetic peptide made to a region surrounding the phosphorylated Serine 1981 of human ATM. [UniProt# Q13315] |
Conjugate | DyLight 650 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: A-T mutated; AT mutated; AT mutated A-T mutated AT1 ATA ataxia telangiectasia mutated (includes complementation groups A C and D) ataxia telangiectasia mutatedATD ATC ATDC ATE DKFZp781A0353 EC 2.7.11.1 MGC74674 serine-protein kinase ATM TEL1 TEL1 telomere maintenance 1 homolog TELO1; Ataxia telangiectasia mutated; Serine-protein kinase ATM; TEL1, telomere maintenance 1, homolog
Gene Aliases: AT1; ATA; ATC; ATD; ATDC; ATE; ATM; TEL1; TELO1
UniProt ID: (Human) Q13315
Entrez Gene ID: (Human) 472
Molecular Function: kinase non-receptor serine/threonine protein kinase nucleic acid binding nucleotide kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.