Abnova
Manufacturer Code:H00000472M05
Catalog # 89-123-139
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2C10 |
Immunogen | ATM (AAH07023.1 1 a.a. ~ 138 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant ATM.
For Research Use Only
Protein Aliases: A-T mutated; AT mutated; AT1; ATA; Ataxia telangiectasia mutated; ATC; ATD; ATDC; ATE; DKFZp781A0353; MGC74674; Serine-protein kinase ATM; TEL1; TEL1, telomere maintenance 1, homolog; TELO1
Gene Aliases: AT1; ATA; ATC; ATD; ATDC; ATE; ATM; TEL1; TELO1
UniProt ID: (Human) Q13315
Entrez Gene ID: (Human) 472
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
nucleic acid binding
nucleotide kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.