R&D Systems
Manufacturer Code:AF2290SP
Catalog # AF2290SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human ATM Arg2138-Arg2400 Accession # Q13315 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: A-T mutated; AT mutated; AT mutated A-T mutated AT1 ATA ataxia telangiectasia mutated (includes complementation groups A C and D) ataxia telangiectasia mutatedATD ATC ATDC ATE DKFZp781A0353 EC 2.7.11.1 MGC74674 serine-protein kinase ATM TEL1 TEL1 telomere maintenance 1 homolog TELO1 TPLL; Ataxia telangiectasia mutated; Serine-protein kinase ATM; TEL1, telomere maintenance 1, homolog
Gene Aliases: AT1; ATA; ATC; ATD; ATDC; ATE; ATM; TEL1; TELO1
UniProt ID: (Human) Q13315
Entrez Gene ID: (Human) 472
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
nucleic acid binding
nucleotide kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.