Abnova
Manufacturer Code:PAB12971
Catalog # 89-116-226
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to C-terminus 14 amino acids of human ATM. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of ATM.
For Research Use Only
Protein Aliases: A-T mutated; AT mutated; AT1; ATA; Ataxia telangiectasia mutated; ATC; ATD; ATDC; ATE; DKFZp781A0353; MGC74674; Serine-protein kinase ATM; TEL1; TEL1, telomere maintenance 1, homolog; TELO1
Gene Aliases: AT1; ATA; ATC; ATD; ATDC; ATE; ATM; TEL1; TELO1
UniProt ID: (Human) Q13315
Entrez Gene ID: (Human) 472
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
nucleic acid binding
nucleotide kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.