Novus Biologicals
Manufacturer Code:NBP201941
Catalog # NBP201941
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1D2 |
Immunogen | Full length human recombinant protein of human ATIC(NP_004035) produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase; 5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase/IMPcyclohydrolase 5-aminoimidazole-4-carboxamide-1-beta-D-ribonucleotidetransformylase/inosinicase AICAR AICAR formyltransferase/IMP cyclohydrolase bifunctional enzyme AICARFT AICARFT/IMPCHASE bifunctional purine biosynthesis protein PURH IMPCHASE phosphoribosylaminoimidazolecarboxamide formyltransferase/IMP cyclohydrolase PURHFLJ93545; 5-aminoimidazole-4-carboxamide-1-beta-D-ribonucleotide transformylase/inosinicase; AICAR formyltransferase; AICAR formyltransferase/IMP cyclohydrolase bifunctional enzyme; AICAR transformylase; AICAR transformylase/inosine monophosphate cyclohydrolase; AICARFT/IMPCHASE; ATIC; Bifunctional purine biosynthesis protein ATIC; Bifunctional purine biosynthesis protein ATIC, N-terminally processed; epididymis secretory sperm binding protein Li 70p; IMP cyclohydrolase; IMP synthase; Inosine 5'-monophosphate cyclohydrolase; Inosinicase; Phosphoribosylaminoimidazolecarboxamide formyltransferase; phosphoribosylaminoimidazolecarboxamide formyltransferase/IMP cyclohydrolase
Gene Aliases: AICAR; AICARFT; ATIC; HEL-S-70p; IMPCHASE; OK/SW-cl.86; PURH
UniProt ID: (Human) P31939
Entrez Gene ID: (Human) 471
Molecular Function:
hydrolase
methyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.