Abnova
Manufacturer Code:MAB7973
Catalog # 89-113-257
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | F38P7H9 |
Immunogen | A synthetic peptide (conjugated with ovalbumin) corresponding to human ATIC. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against synthetic peptide of ATIC.
For Research Use Only
Protein Aliases: 5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase; 5-aminoimidazole-4-carboxamide-1-beta-D-ribonucleotide transformylase/inosinicase; AICAR; AICAR formyltransferase; AICAR formyltransferase/IMP cyclohydrolase bifunctional enzyme; AICAR transformylase; AICAR transformylase/inosine monophosphate cyclohydrolase; AICARFT; AICARFT/IMPCHASE; ATIC; Bifunctional purine biosynthesis protein ATIC; Bifunctional purine biosynthesis protein ATIC, N-terminally processed; epididymis secretory sperm binding protein Li 70p; FLJ93545; IMP cyclohydrolase; IMP synthase; IMPCHASE; Inosine 5'-monophosphate cyclohydrolase; Inosinicase; Phosphoribosylaminoimidazolecarboxamide formyltransferase; phosphoribosylaminoimidazolecarboxamide formyltransferase/IMP cyclohydrolase; PURH
Gene Aliases: AICAR; AICARFT; ATIC; HEL-S-70p; IMPCHASE; OK/SW-cl.86; PURH
UniProt ID: (Human) P31939
Entrez Gene ID: (Human) 471
Molecular Function: hydrolase methyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.