Invitrogen
Manufacturer Code:PA521469
Catalog # PIPA521469
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 310 and 580 of ATIC (Uniprot IDNo.P31939) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase; 5-aminoimidazole-4-carboxamide-1-beta-D-ribonucleotide transformylase/inosinicase; AICAR formyltransferase; AICAR formyltransferase/IMP cyclohydrolase bifunctional enzyme; AICAR transformylase; AICAR transformylase/inosine monophosphate cyclohydrolase; AICARFT/IMPCHASE; ATIC; Bifunctional purine biosynthesis protein ATIC; Bifunctional purine biosynthesis protein ATIC, N-terminally processed; epididymis secretory sperm binding protein Li 70p; IMP cyclohydrolase; IMP synthase; Inosine 5'-monophosphate cyclohydrolase; Inosinicase; OK/SW-cl.86 AICAR AICARFT IMPCHASE PURH 5-aminoimidazole-4-carboxamide-1-beta-D-ribonucleotide transformylase/inosinicase AICAR formyltransferase/IMP cyclohydrolase bifunctional enzyme AICARFT/IMPCHASE bifunctional purine biosynthesis protein PURH phosphoribosylaminoimidazolecarboxamide formyltransferase/IMP cyclohydrolase; Phosphoribosylaminoimidazolecarboxamide formyltransferase; phosphoribosylaminoimidazolecarboxamide formyltransferase/IMP cyclohydrolase
Gene Aliases: AICAR; AICARFT; ATIC; HEL-S-70p; IMPCHASE; OK/SW-cl.86; PURH
UniProt ID: (Human) P31939
Entrez Gene ID: (Human) 471
Molecular Function:
hydrolase
methyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.