Invitrogen
Manufacturer Code:PA576871
Catalog # PIPA576871
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human ATGL. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adipose triglyceride lipase; Adipose triglyceride lipaseATGLATGL DESNUTRINCalcium independent phospholipase A2Calcium-independent phospholipase A2DesnutrinFP17548IPLA2 zetaIPLA2-zetaPATATIN LIKE PHOSPHOLIPASE DOMAIN CONTAINING PROTEIN 2Patatin-like phospholipase domain-containing protein 2PEDF RPHOSPHOLIPASE A2 CALCIUM INDEPENDENT ZETAPigment epithelium derived factorPigment epithelium-derived factorplplplpl2PLPL2&lowbarHUMANPnpla2Transport secretion protein 2Transport secretion protein 2.2Transport-secretion protein 2Triglyceride hydrolaseTTS 2.2TTS2TTS2.2ZETA; Calcium-independent phospholipase A2; Desnutrin; IPLA2-zeta; mutant patatin-like phospholipase domain containing 2; patatin-like phospholipase domain containing 2; patatin-like phospholipase domain containing protein 2; Patatin-like phospholipase domain-containing protein 2; Pigment epithelium-derived factor; transport-secretion protein; Transport-secretion protein 2; transport-secretion protein 2.2; triglyceride hydrolase; TTS2; TTS2.2
Gene Aliases: 0610039C21Rik; 1110001C14Rik; ATGL; FP17548; iPLA2zeta; PEDF-R; PNPLA2; RGD1309044; TTS-2.2; TTS2
UniProt ID: (Human) Q96AD5, (Rat) P0C548, (Mouse) Q8BJ56
Entrez Gene ID: (Human) 57104, (Rat) 361676, (Mouse) 66853
Molecular Function: acyltransferase hydrolase lipase phospholipase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.