Invitrogen
Manufacturer Code:PA517436
Catalog # PIPA517436
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a sequence around Pro186 of human ATGL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adipose triglyceride lipase; Desnutrin; IPLA2-zeta TTS2.2 adipose triglyceride lipase calcium-independent phospholipase A2 desnutrin patatin-like phospholipase domain-containing protein 2 pigment epithelium-derived factor transport-secretion protein 2.2 triglyceride hydrolase; Patatin-like phospholipase domain-containing protein 2; transport-secretion protein; triglyceride hydrolase
Gene Aliases: 0610039C21Rik; 1110001C14Rik; Atgl; Pnpla2; RGD1309044; TTS-2.2
UniProt ID: (Rat) P0C548, (Mouse) Q8BJ56
Entrez Gene ID: (Rat) 361676, (Mouse) 66853
Molecular Function: acyltransferase hydrolase lipase phospholipase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.