Invitrogen
Manufacturer Code:PA5110189
Catalog # PIPA5110189
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 265-504 of human PNPLA2 (NP_065109.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 0610039C21Rik 1110001C14Rik Adipose triglyceride lipase Atgl ATGL DESNUTRIN Calcium-independent phospholipase A2 Desnutrin DKFZp667M109 FP17548 IPLA2 zeta iPLA2zeta IPLA2-zeta Patatin-like phospholipase domain-containing protein 2 PEDF R PEDF-R Pigment epithelium-derived factor plpl plpl2 PNPLA2 RGD1309044 Transport-secretion protein 2 TTS 2.2 TTS2 TTS2.2 TTS-2.2; Adipose triglyceride lipase; Calcium-independent phospholipase A2; Desnutrin; IPLA2-zeta; mutant patatin-like phospholipase domain containing 2; patatin-like phospholipase domain containing 2; patatin-like phospholipase domain containing protein 2; Patatin-like phospholipase domain-containing protein 2; Pigment epithelium-derived factor; Transport-secretion protein 2; transport-secretion protein 2.2; triglyceride hydrolase; TTS2; TTS2.2
Gene Aliases: 1110001C14Rik; ATGL; FP17548; iPLA2zeta; PEDF-R; PNPLA2; TTS-2.2; TTS2
UniProt ID: (Human) Q96AD5
Entrez Gene ID: (Human) 57104
Molecular Function: acyltransferase hydrolase lipase phospholipase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.