Novus Biologicals
Manufacturer Code:NBP267616
Catalog # NBP267616
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | SC67-05 |
Immunogen | Recombinant Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: APG9 autophagy 9-like 1; APG9 autophagy 9-like 1 APG9 autophagy 9-like 1 (S. cerevisiae) APG9L1APG9-like 1 ATG9 autophagy related 9 homolog A (S. cerevisiae) autophagy 9-like 1 protein autophagy-related protein 9A FLJ22169 mATG9 MGD3208; APG9-like 1; ATG9 autophagy related 9 homolog A; autophagy 9-like 1 protein; Autophagy-related protein 9A; mATG9
Gene Aliases: APG9L1; ATG9A; mATG9; MGD3208
UniProt ID: (Human) Q7Z3C6
Entrez Gene ID: (Human) 79065
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.