R&D Systems
Manufacturer Code:FAB6608S100UG
Catalog # FAB6608S100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 683906 |
Immunogen | <EM>E. coli</EM>-derived recombinant human ATG7 Asn387-Gln570 Accession # O95352 |
Conjugate | Alexa Fluor 750 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: APG7 APG7 autophagy 7-like (S. cerevisiae) APG7L APG7-LIKE ATG12-activating enzyme E1 ATG7 ATG7 autophagy related 7 homolog (S. cerevisiae) Autophagy-related protein 7 DKFZp434N0735 GSA7 hAGP7 ubiquitin activating enzyme E1-like protein Ubiquitin-activating enzyme E1-like protein ubiquitin-like modifier-activating enzyme ATG7; APG7-like; ATG12-activating enzyme E1 ATG7; Autophagy-related protein 7; hAGP7; Ubiquitin-activating enzyme E1-like protein; Ubiquitin-like modifier-activating enzyme ATG7
Gene Aliases: APG7-LIKE; APG7L; ATG7; GSA7
UniProt ID: (Human) O95352
Entrez Gene ID: (Human) 10533
Molecular Function:
ligase
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.