Invitrogen
Manufacturer Code:PA520174
Catalog # PIPA520174
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 17 amino acid peptide from near the amino terminus of human APG7. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: APG7-like; APG7-LIKE APG7L GSA7 ATG12-activating enzyme E1 ATG7 autophagy-related protein 7 hAGP7 ubiquitin activating enzyme E1-like protein ubiquitin-activating enzyme E1-like protein ubiquitin-like modifier-activating enzyme ATG7; ATG12-activating enzyme E1 ATG7; Autophagy-related protein 7; hAGP7; Ubiquitin-activating enzyme E1-like protein; Ubiquitin-like modifier-activating enzyme ATG7
Gene Aliases: 1810013K23Rik; Agp7; APG7-LIKE; APG7L; ATG7; Atg7l; Gm21553; GSA7
UniProt ID: (Human) O95352, (Mouse) Q9D906
Entrez Gene ID: (Human) 10533, (Mouse) 74244
Molecular Function:
ligase
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.