Novus Biologicals
Manufacturer Code:NBP176650
Catalog # NBP176650
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a 17 amino acid synthetic peptide from near the amino terminus of human APG7. The immunogen is located within the first 50 amino acids of APG7. Amino Acid Squence: QKKLNEYRLDEAPKDIK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: APG7 autophagy 7-like (S. cerevisiae) APG7L APG7-LIKE ATG12-activating enzyme E1 ATG7 ATG7 autophagy related 7 homolog (S. cerevisiae) Autophagy-related protein 7 DKFZp434N0735 GSA7 hAGP7 ubiquitin activating enzyme E1-like protein Ubiquitin-activating enzyme E1-like protein ubiquitin-like modifier-activating enzyme ATG7; APG7-like; ATG12-activating enzyme E1 ATG7; Autophagy-related protein 7; hAGP7; Ubiquitin-activating enzyme E1-like protein; Ubiquitin-like modifier-activating enzyme ATG7
Gene Aliases: APG7-LIKE; APG7L; ATG7; GSA7
UniProt ID: (Human) O95352
Entrez Gene ID: (Human) 10533
Molecular Function:
ligase
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.