Abnova
Manufacturer Code:PAB13170
Catalog # 89-116-418
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to N-terminus 17 amino acids of human ATG7. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of ATG7.
For Research Use Only
Protein Aliases: APG7-LIKE; APG7L; ATG12-activating enzyme E1 ATG7; Autophagy-related protein 7; DKFZp434N0735; GSA7; hAGP7; Ubiquitin-activating enzyme E1-like protein; Ubiquitin-like modifier-activating enzyme ATG7
Gene Aliases: APG7-LIKE; APG7L; ATG7; GSA7
UniProt ID: (Human) O95352
Entrez Gene ID: (Human) 10533
Molecular Function: ligase transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.