R&D Systems
Manufacturer Code:MAB5294SP
Catalog # MAB5294SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 603813 |
Immunogen | <EM>E. coli</EM>-derived recombinant human ATG5 Asn99-Thr193 Accession # Q9H1Y0 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: APG5 APG5 autophagy 5-like (S. cerevisiae) ASP ASPAPG5-LIKE ATG5 autophagy related 5 homolog (S. cerevisiae) Autophagy protein 5; APG5-like; Apoptosis-specific protein; ATG5 autophagy related 5 homolog; Autophagy protein 5
Gene Aliases: APG5; APG5-LIKE; APG5L; ASP; ATG5; hAPG5
UniProt ID: (Human) Q9H1Y0
Entrez Gene ID: (Human) 9474
Molecular Function: membrane traffic protein membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.