Invitrogen
Manufacturer Code:PA523186
Catalog # PIPA523186
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide (19 CFTLYQDEITEREAEP 34) of APG5L protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: APG5 APG5-LIKE APG5L ASP hAPG5 apoptosis specific protein apoptosis-specific protein autophagy protein 5; APG5-like; Apoptosis-specific protein; ATG5 autophagy related 5 homolog; Autophagy protein 5; autophagy-related 5
Gene Aliases: 2010107M05Rik; 3110067M24Rik; APG5; APG5-LIKE; APG5L; ASP; ATG5; Atg5l; AW319544; C88337; hAPG5; Paddy
UniProt ID: (Human) Q9H1Y0, (Bovine) Q3MQ24, (Mouse) Q99J83
Entrez Gene ID: (Human) 9474, (Bovine) 532686, (Mouse) 11793, (Rat) 365601
Molecular Function:
membrane traffic protein
membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.