Novus Biologicals
Manufacturer Code:NBP280585
Catalog # NB013658
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Baker's yeast |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 7C2NB |
Immunogen | Partial recombinant protein made to an internal sequence of the S. cerevisiae ATG11 protein (between amino acids 50-250) [UniProt Q12527] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: autophagy protein ATG11; Autophagy-related protein 11; Cytoplasm to vacuole targeting protein 9
Gene Aliases: ATG11; CVT3; CVT9; YP9499.07c; YPR049C
UniProt ID: (Baker's yeast) Q12527
Entrez Gene ID: (Baker's yeast) 856162
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.