Novus Biologicals
Manufacturer Code:NBP1402560.1MG
Catalog # NBP14025601
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 70B1413.1 |
Immunogen | This monoclonal antibody was made against a partial protein containing amino acids 1-273 of human ATF6. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: activating transcription factor 6 Activating transcription factor 6 alpha atf6 a ATF6A ATF6-alpha cAMP-dependent transcription factor ATF-6 alpha cyclic AMP-dependent transcription factor ATF-6 alpha; Activating transcription factor 6 alpha; ATF6-alpha; cAMP-dependent transcription factor ATF-6 alpha; Cyclic AMP-dependent transcription factor ATF-6 alpha; Processed cyclic AMP-dependent transcription factor ATF-6 alpha
Gene Aliases: ACHM7; ATF6; ATF6A
UniProt ID: (Human) P18850
Entrez Gene ID: (Human) 22926
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.