R&D Systems
Manufacturer Code:MAB71527100
Catalog # MAB71527100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 2358C |
Immunogen | <em>E. coli</em>-derived recombinant human ATF6 Met1-Thr192 Accession No. P18850 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 °C as supplied. 1 month 2 to 8 °C under sterile conditions after reconstitution. 6 months -20 to -70 °C under sterile condition |
Rabbit Monoclonal Antibody
Protein Aliases: ACHM7 activating transcription factor 6 Activating transcription factor 6 alpha atf6 a ATF6 alpha ATF6A ATF6-alpha cAMP-dependent transcription factor ATF-6 alpha cyclic AMP-dependent transcription factor ATF-6 alpha; Activating transcription factor 6 alpha; ATF6-alpha; cAMP-dependent transcription factor ATF-6 alpha; Cyclic AMP-dependent transcription factor ATF-6 alpha; Processed cyclic AMP-dependent transcription factor ATF-6 alpha
Gene Aliases: ACHM7; ATF6; ATF6A
UniProt ID: (Human) P18850
Entrez Gene ID: (Human) 22926
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.