Invitrogen
Manufacturer Code:PA520216
Catalog # PIPA520216
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 17 amino acid peptide from near the amino terminus of human ATF6. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Activating transcription factor 6 alpha; ATF6-alpha; ATF6A cAMP-dependent transcription factor ATF-6 alpha cyclic AMP-dependent transcription factor ATF-6 alpha; cAMP-dependent transcription factor ATF-6 alpha; Cyclic AMP-dependent transcription factor ATF-6 alpha; Processed cyclic AMP-dependent transcription factor ATF-6 alpha
Gene Aliases: 9130025P16Rik; 9630036G24; AA789574; ACHM7; ATF6; ATF6A; Atf6alpha; ESTM49
UniProt ID: (Human) P18850, (Mouse) F6VAN0
Entrez Gene ID: (Human) 22926, (Mouse) 226641
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.