Invitrogen
Manufacturer Code:MA538080
Catalog # PIMA538080
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC2043 |
Immunogen | A synthesized peptide derived from human ATF5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: Activating transcription factor 5; Activating transcription factor 5 activating transcription factor 5-alpha/beta activating transcription factor 7 activating transcription factor X AFTA ATF5 Atf7 Atfx BZIP protein ATF7 cAMP-dependent transcription factor ATF-5 cyclic AMP-dependent transcription factor ATF-5 FLJ34666 HMFN0395 NAP1 NRIF3-associated protein ODA-10 Transcription factor ATFx transcription factor-like protein ODA-10; Activating transcription factor 5-alpha/beta; activating transcription factor 7; activating transcription factor X; BZIP protein ATF7; cAMP-dependent transcription factor ATF-5; Cyclic AMP-dependent transcription factor ATF-5; NAP1; NRIF3-associated protein; Transcription factor ATFx; Transcription factor-like protein ODA-10
Gene Aliases: AFTA; ATF5; Atf7; ATFX; HMFN0395; Nap1; ODA-10
UniProt ID: (Human) Q9Y2D1, (Rat) Q6P788, (Mouse) O70191
Entrez Gene ID: (Human) 22809, (Rat) 282840, (Mouse) 107503
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.