Invitrogen
Manufacturer Code:PA5114124
Catalog # PIPA5114124
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal region of Rat Atf5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Activating transcription factor 5; Activating transcription factor 5 Activating transcription factor 5-alpha/beta activating transcription factor 7 activating transcription factor X AFTA Atf5 Atf7 ATFX BZIP protein ATF7 cAMP-dependent transcription factor ATF-5 Cyclic AMP-dependent transcription factor ATF-5 FLJ34666 HMFN0395 Nap1 NRIF3-associated protein ODA-10 Transcription factor ATFx Transcription factor-like protein ODA-10; cAMP-dependent transcription factor ATF-5; Cyclic AMP-dependent transcription factor ATF-5; Transcription factor ATFx
Gene Aliases: Atf5; Atfx
UniProt ID: (Rat) Q6P788
Entrez Gene ID: (Rat) 282840
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.