Invitrogen
Manufacturer Code:PA568802
Catalog # PIPA568802
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of mouse ATF4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Activating transcription factor 4; activating transcription factor 4 (tax-responsive enhancer element B67); activating transcription factor ATF-4; atf4 {ECO:0000312|RGD:621863}; C/ATF; C/EBP-related ATF; cAMP response element-binding protein 2; cAMP-dependent transcription factor ATF-4; cAMP-responsive element-binding protein 2; CREB-2; CREB-2 CREB2 TAXREB67 TXREB DNA-binding protein TAXREB67 cAMP response element-binding protein 2 cAMP-dependent transcription factor ATF-4 cAMP-responsive element-binding protein 2 cyclic AMP-dependent transcription factor ATF-4 cyclic AMP-responsive element-binding protein 2 tax-responsive enhancer element-binding protein 67; Cyclic AMP-dependent transcription factor ATF-4; Cyclic AMP-responsive element-binding protein 2; DNA-binding protein TAXREB67; rATF-4; tax-responsive enhancer element B67; Tax-responsive enhancer element-binding protein 67; tax-responsive enhancer element-binding protein 67 homolog; TaxREB67; taxREB67 homolog
Gene Aliases: Atf-4; ATF4; C/ATF; CREB-2; CREB2; TAXREB67; TXREB
UniProt ID: (Human) P18848, (Bovine) Q3ZCH6, (Rat) Q9ES19, (Mouse) Q06507
Entrez Gene ID: (Horse) 100070334, (Dog) 474503, (Sheep) 100216465, (Human) 468, (Bovine) 509107, (Rat) 79255, (Guinea pig) 100732296, (Mouse) 11911
Molecular Function: CREB transcription factor basic leucine zipper transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.