Novus Biologicals
Manufacturer Code:NBP152647
Catalog # NBP152647
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen for this product maps to a region between residue 221 and 271 of human Activating Transcription Factor 1 using the numbering given in entry NP_005162.1 (GeneID 466). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Activating transcription factor 1; activating transcription factor 1FUS/ATF-1 cAMP-dependent transcription factor ATF-1 EWS-ATF1 Protein TREB36 TREB36cyclic AMP-dependent transcription factor ATF-1; cAMP-dependent transcription factor ATF-1; Cyclic AMP-dependent transcription factor ATF-1; Protein TREB36
Gene Aliases: ATF1; EWS-ATF1; FUS/ATF-1; TREB36
UniProt ID: (Human) P18846
Entrez Gene ID: (Human) 466
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.