Novus Biologicals
Manufacturer Code:NBP234126
Catalog # NBP234126
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: KAQVQVPQQSHQQILPQQQQNQLSIAQSHSALLQPSQHPEKKNKLVIKEKEKESQRERDSAEGGEGNTGPKETLPDA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha-fetoprotein enhancer binding protein; Alpha-fetoprotein enhancer-binding protein; Alpha-fetoprotein enhancer-binding protein AT motif-binding factor AT motif-binding factor 1 ATBF1alpha-fetoprotein enhancer binding protein AT-binding transcription factor 1ATBT ZFH-3 zinc finger homeobox 3 zinc finger homeobox protein 3 Zinc finger homeodomain protein 3 ZNF927; AT motif-binding factor 1; AT-binding transcription factor 1; ZFH-3; Zinc finger homeobox protein 3; Zinc finger homeodomain protein 3
Gene Aliases: ATBF1; ATBT; C16orf47; ZFHX3; ZNF927
UniProt ID: (Human) Q15911
Entrez Gene ID: (Human) 463
Molecular Function: RNA binding protein actin family cytoskeletal protein cytoskeletal protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.