R&D Systems
Manufacturer Code:AF7384
Catalog # AF7384
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human ATBF1/ZFHX3 His3608-Leu3703 Accession # Q15911 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha-fetoprotein enhancer binding protein; Alpha-fetoprotein enhancer-binding protein; Alpha-fetoprotein enhancer-binding protein AT motif-binding factor AT motif-binding factor 1 ATBF1alpha-fetoprotein enhancer binding protein AT-binding transcription factor 1ATBT ATBT ZFH3 ZFH-3 ZFHX3 zinc finger homeobox 3 zinc finger homeobox protein 3 Zinc finger homeodomain protein 3 ZNF927; AT motif-binding factor 1; AT-binding transcription factor 1; ZFH-3; Zinc finger homeobox protein 3; Zinc finger homeodomain protein 3
Gene Aliases: ATBF1; ATBT; C16orf47; ZFHX3; ZNF927
UniProt ID: (Human) Q15911
Entrez Gene ID: (Human) 463
Molecular Function: RNA binding protein actin family cytoskeletal protein cytoskeletal protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.