Invitrogen
Manufacturer Code:PA569187
Catalog # PIPA569187
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human ATAT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acetyltransferase mec-17 homolog; acetyltransferase mec-17 homolog {ECO:0000255|HAMAP-Rule:MF_03130}; Alpha-TAT; alpha-TAT {ECO:0000255|HAMAP-Rule:MF_03130}; alpha-TAT1; alpha-TAT1 {ECO:0000255|HAMAP-Rule:MF_03130}; Alpha-tubulin N-acetyltransferase 1; alpha-tubulin N-acetyltransferase 1 {ECO:0000255|HAMAP-Rule:MF_03130}; Novel DUF738 containing protein (2610110G12Rik); TAT; TAT {ECO:0000255|HAMAP-Rule:MF_03130}
Gene Aliases: 0610011P08Rik; 2610008K08Rik; 2610110G12Rik; 3110080J08Rik; alpha-TAT; alpha-TAT1; ATAT1; C23H6orf134; C6orf134; MEC17; Nbla00487; RGD1303066; TAT
UniProt ID: (Human) Q5SQI0, (Mouse) Q8K341, (Rat) Q6MG11
Entrez Gene ID: (Rabbit) 100358006, (Dog) 477580, (Horse) 100055595, (Bovine) 786491, (Human) 79969, (Mouse) 73242, (Rat) 361789, (Guinea pig) 100719818
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.