Invitrogen
Manufacturer Code:PA5112992
Catalog # PIPA5112992
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Alpha-tubulin N-acetyltransferase 1 protein (194-238AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 0610011P08Rik 2610008K08Rik 2610110G12Rik 3110080J08Rik Acetyltransferase mec-17 homolog acetyltransferase mec-17 homolog {ECO:0000255 alpha tubulin acetyltransferase 1 Alpha-TAT alpha-TAT {ECO:0000255 alpha-TAT1 alpha-TAT1 {ECO:0000255 alpha-tubulin N-acetyltransferase Alpha-tubulin N-acetyltransferase 1 alpha-tubulin N-acetyltransferase 1 {ECO:0000255 Atat1 BOS_21654 C23H6orf134 C6orf134 HAMAP-Rule:MF_03130} LOC100358006 Mec17 Nbla00487 Novel DUF738 containing protein (2610110G12Rik) RGD1303066 TAT TAT {ECO:0000255 UPF0635 protein C6orf134-like; Acetyltransferase mec-17 homolog; acetyltransferase mec-17 homolog {ECO:0000255|HAMAP-Rule:MF_03130}; Alpha-TAT; alpha-TAT {ECO:0000255|HAMAP-Rule:MF_03130}; alpha-TAT1; alpha-TAT1 {ECO:0000255|HAMAP-Rule:MF_03130}; Alpha-tubulin N-acetyltransferase 1; alpha-tubulin N-acetyltransferase 1 {ECO:0000255|HAMAP-Rule:MF_03130}; Novel DUF738 containing protein (2610110G12Rik); TAT; TAT {ECO:0000255|HAMAP-Rule:MF_03130}
Gene Aliases: 0610011P08Rik; 2610008K08Rik; 2610110G12Rik; 3110080J08Rik; alpha-TAT; alpha-TAT1; ATAT1; C6orf134; MEC17; Nbla00487; RGD1303066; TAT
UniProt ID: (Human) Q5SQI0, (Mouse) Q8K341, (Rat) Q6MG11
Entrez Gene ID: (Human) 79969, (Mouse) 73242, (Rat) 361789
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.