Invitrogen
Manufacturer Code:PA546087
Catalog # PIPA546087
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human ATAD2B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATPase family AAA domain containing 2B; ATPase family AAA domain-containing protein 2B
Gene Aliases: 1110014E10Rik; ATAD2B; BC032887; C79189; D530031C13Rik; KIAA1240
UniProt ID: (Human) Q9ULI0
Entrez Gene ID: (Human) 54454, (Horse) 100071677, (Dog) 475681, (Rabbit) 100340219, (Bovine) 518785, (Mouse) 320817
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.