Novus Biologicals
Manufacturer Code:NBP214786
Catalog # NBP214786
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CL0182 |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: TAYAIIKEELDEDFEQLCEEIQESRKKRGCSSSKYAPSYYHVMPKQNSTLVGDKRSDPEQNEKLKTPSTPVACSTPAQLKRKIRKKSNWYLGTIKKRRKISQAKDDSQNAIDHK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AAA nuclear coregulator cancer-associated protein; AAA nuclear coregulator cancer-associated protein ANCCAMGC131938 ATPase family AAA domain-containing protein 2 ATPase family AAA domain containing 2 DKFZp667N1320 EC 3.6.1.3 MGC142216 MGC29843 MGC5254 PRO2000; ANCCA; ATPase family AAA domain-containing protein 2
Gene Aliases: ANCCA; ATAD2; CT137; L16; PRO2000
UniProt ID: (Human) Q6PL18
Entrez Gene ID: (Human) 29028
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.