Proteintech
Manufacturer Code:238941AP150UL
Catalog # 50-173-4154
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ATAD2 Fusion Protein Ag20947 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AAA nuclear coregulator cancer-associated protein; ANCCA; ANCCA ATAD2 PRO2000; ATPase family AAA domain-containing protein 2
Gene Aliases: ANCCA; ATAD2; CT137; L16; PRO2000
UniProt ID: (Human) Q6PL18
Entrez Gene ID: (Human) 29028
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.