Invitrogen
Manufacturer Code:PA5113230
Catalog # PIPA5113230
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Ankyrin repeat SAM and basic leucine zipper domain-containing protein 1 protein (1-200AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4933400N19Rik ALP1 ANKL1 ankyrin repeat SAM and basic leucine zipper domain containing 1 Ankyrin repeat SAM and basic leucine zipper domain-containing protein 1 ankyrin-like 1 Ankyrin-like protein 1 ASZ1 C7orf7 CT1.19 Gasz germ cell-specific ankyrin SAM and basic leucine zipper domain containing protein Germ cell-specific ankyrin SAM and basic leucine zipper domain-containing protein ORF3; Ankyrin repeat, SAM and basic leucine zipper domain-containing protein 1; ankyrin-like 1; Ankyrin-like protein 1; ASZ1; Germ cell-specific ankyrin, SAM and basic leucine zipper domain-containing protein
Gene Aliases: ALP1; ANKL1; ASZ1; C7orf7; CT1.19; GASZ; Orf3
UniProt ID: (Human) Q8WWH4
Entrez Gene ID: (Human) 136991, (Rat) 170578
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.