Invitrogen
Manufacturer Code:PA570292
Catalog # PIPA570292
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human ASXL2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: additional sex combs like 2; additional sex combs like 2, transcriptional regulator; additional sex combs like transcriptional regulator 2; Additional sex combs-like protein 2; ASXH2 KIAA1685; polycomb group protein ASXH2; polycomb group protein ASXH2 homolog; Putative Polycomb group protein ASXL2
Gene Aliases: 4930556B16Rik; ASXH2; ASXL2; KIAA1685; mKIAA1685
UniProt ID: (Human) Q76L83, (Mouse) Q8BZ32
Entrez Gene ID: (Horse) 100071411, (Rabbit) 100355851, (Dog) 100684260, (Pig) 100515136, (Human) 55252, (Guinea pig) 100719241, (Mouse) 75302, (Rat) 313922
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.